Assay Details
| Gene Symbol: CA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178572.2 | NP_848667.1 | 6 - 7 | 711 | 69 | |
| GenBank mRNA | AY240020.1 | - | 6 - 7 | 711 | 69 | |
| BC103260.1 | - | 6 - 7 | 765 | 69 | ||
More Information
Set Membership: |
|

CA2
Genomic Map