Assay Details
| Gene Symbol: CAPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178574.3 | NP_848669.1 | 7 - 8 | 858 | 58 | |
| GenBank mRNA | AY219899.1 | - | 7 - 8 | 859 | 58 | |
| BC102160.1 | - | 7 - 8 | 829 | 58 | ||
More Information
Set Membership: |
|

CAPG
Genomic Map