Assay Details
| Gene Symbol: TRDMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181812.2 | NP_861528.1 | 3 - 4 | 270 | 73 | |
| GenBank mRNA | AY244708.1 | - | 3 - 4 | 273 | 73 | |
| BT021768.1 | - | 3 - 4 | 266 | 73 | ||
More Information
Set Membership: |
|

TRDMT1
Genomic Map