Assay Details
| Gene Symbol: RAX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182653.1 | NP_872594.1 | 2 - 3 | 263 | 64 | |
| GenBank mRNA | AY211276.1 | - | 2 - 3 | 263 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAX2| Gene Symbol: RAX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182653.1 | NP_872594.1 | 2 - 3 | 263 | 64 | |
| GenBank mRNA | AY211276.1 | - | 2 - 3 | 263 | 64 | |
Set Membership: |
|