Assay Details
| Gene Symbol: TFPI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182788.1 | NP_877589.1 | 4 - 5 | 690 | 160 | |
| GenBank mRNA | AY234861.1 | - | 4 - 5 | 690 | 160 | |
| BC103205.1 | - | 4 - 5 | 699 | 160 | ||
More Information
Set Membership: |
|

TFPI2
Genomic Map