Assay Details
| Gene Symbol: RDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183363.1 | NP_899207.1 | 5 - 6 | 661 | 52 | |
| GenBank mRNA | AY115489.1 | - | 5 - 6 | 661 | 52 | |
| BC142243.1 | - | 5 - 6 | 845 | 52 | ||
More Information
Set Membership: |
|

RDH12
Genomic Map