Assay Details
| Gene Symbol: SPG21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205772.1 | NP_991341.1 | 6 - 7 | 688 | 89 | |
| GenBank mRNA | AF451182.1 | - | 6 - 7 | 688 | 89 | |
| BC118320.1 | - | 6 - 7 | 740 | 89 | ||
More Information
Set Membership: |
|

SPG21
Genomic Map