Assay Details
| Gene Symbol: SLCO1B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205804.2 | NP_991373.1 | 10 - 11 | 1564 | 88 | |
| GenBank mRNA | AJ508747.1 | - | 10 - 11 | 1568 | 88 | |
More Information
Set Membership: |
|

SLCO1B3
Genomic Map