Assay Details
| Gene Symbol: HNF4G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001359925.1 | NP_001346854.1 | 7 - 8 | 988 | 79 | |
| NM_205811.3 | NP_991380.2 | 6 - 7 | 791 | 79 | ||
| GenBank mRNA | AY318753.1 | - | 6 - 7 | 79 | ||
| BC151653.1 | - | 7 - 8 | 79 | |||
More Information
Set Membership: |
|

HNF4G
Genomic Map