Assay Details
| Gene Symbol: SERPINA3-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012283.3 | NP_001012283.2 | 3 - 4 | 963 | 74 | |
| GenBank mRNA | AY386699.1 | - | 3 - 4 | 926 | 74 | |
| BC102746.1 | - | 3 - 4 | 964 | 74 | ||
| U13609.1 | - | 2 - 3 | 900 | 74 | ||
More Information
Set Membership: |
|

SERPINA3-7
Genomic Map