Assay Details
| Gene Symbol: MYD88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014382.2 | NP_001014404.1 | 4 - 5 | 776 | 114 | |
| GenBank mRNA | AJ853453.1 | - | 4 - 5 | 743 | 114 | |
| AY634627.1 | - | 2 - 3 | 126 | 114 | ||
| AY934808.2 | - | 3 - 4 | 618 | 114 | ||
| AY937381.1 | - | 2 - 3 | 779 | 114 | ||
| BC102851.1 | - | 4 - 5 | 776 | 114 | ||
| KT601039.1 | - | 4 - 5 | 739 | 114 | ||
More Information
Set Membership: |
|

MYD88
Genomic Map