Assay Details
| Gene Symbol: TMBIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014914.2 | NP_001014914.1 | 3 - 4 | 380 | 63 | |
| GenBank mRNA | BC105247.1 | - | 3 - 4 | 380 | 63 | |
| BC151432.1 | - | 3 - 4 | 399 | 63 | ||
| BT021506.1 | - | 3 - 4 | 387 | 63 | ||
More Information
Set Membership: |
|

TMBIM4
Genomic Map