Assay Details
| Gene Symbol: RPL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024469.3 | NP_001019640.2 | 1 | 608 | 87 | |
| GenBank mRNA | AY911321.1 | - | 7 - 8 | 614 | 87 | |
| BC103427.1 | - | 1 | 609 | 87 | ||
More Information
Set Membership: |
|

RPL9
Genomic Map