Assay Details
| Gene Symbol: LPCAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024546.2 | NP_001019717.2 | 2 - 3 | 318 | 59 | |
| GenBank mRNA | BC102802.1 | - | 2 - 3 | 318 | 59 | |
| BT021868.1 | - | 2 - 3 | 289 | 59 | ||
More Information
Set Membership: |
|

LPCAT3
Genomic Map