Assay Details
| Gene Symbol: SRGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025326.2 | NP_001020497.1 | 1 - 2 | 129 | 65 | |
| GenBank mRNA | AY911316.1 | - | 1 - 2 | 129 | 65 | |
| BC102219.1 | - | 1 - 2 | 129 | 65 | ||
More Information
Set Membership: |
|

SRGN
Genomic Map