Assay Details
| Gene Symbol: KRT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033610.1 | NP_001028782.1 | 5 - 6 | 957 | 58 | |
| GenBank mRNA | BC103339.1 | - | 5 - 6 | 957 | 58 | |
| K03532.1 | - | 5 - 6 | 551 | 58 | ||
| X12877.1 | - | 551 | 58 | |||
More Information
Set Membership: |
|

KRT8
Genomic Map