Assay Details
| Gene Symbol: RPS18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033614.2 | NP_001028786.1 | 2 - 3 | 147 | 74 | |
| GenBank mRNA | AB098889.1 | - | 2 - 3 | 128 | 74 | |
| AB098899.1 | - | 1 - 2 | 88 | 74 | ||
| AB099024.1 | - | 2 - 3 | 131 | 74 | ||
| BC102293.1 | - | 2 - 3 | 148 | 74 | ||
More Information
Set Membership: |
|

RPS18
Genomic Map