Assay Details
| Gene Symbol: CTSS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033615.2 | NP_001028787.1 | 7 - 8 | 963 | 74 | |
| GenBank mRNA | BC102245.1 | - | 7 - 8 | 964 | 74 | |
| M95211.1 | - | 3 - 4 | 493 | 74 | ||
| X62001.1 | - | 493 | 74 | |||
More Information
Set Membership: |
|

CTSS
Genomic Map