Assay Details
| Gene Symbol: NOP56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033622.2 | NP_001028794.1 | 3 - 4 | 235 | 77 | |
| GenBank mRNA | BC103114.1 | - | 3 - 4 | 236 | 77 | |
More Information
Set Membership: |
|

NOP56
Genomic Map