Assay Details
| Gene Symbol: PEBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033623.2 | NP_001028795.1 | 2 - 3 | 340 | 79 | |
| GenBank mRNA | BC102389.1 | - | 2 - 3 | 342 | 79 | |
More Information
Set Membership: |
|

PEBP1
Genomic Map