Assay Details
| Gene Symbol: RPS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033624.2 | NP_001028796.1 | 3 - 4 | 298 | 87 | |
| GenBank mRNA | BC102215.1 | - | 3 - 4 | 299 | 87 | |
More Information
Set Membership: |
|

RPS16
Genomic Map