Assay Details
| Gene Symbol: DNAJB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033763.1 | NP_001028935.1 | 1 - 2 | 247 | 102 | |
| GenBank mRNA | BC104503.1 | - | 1 - 2 | 247 | 102 | |
More Information
Set Membership: |
|

DNAJB1
Genomic Map