Assay Details
| Gene Symbol: CD3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034033.2 | NP_001029205.1 | 1 - 2 | 91 | 60 | |
| GenBank mRNA | BC102857.1 | - | 1 - 2 | 92 | 60 | |
| X53269.1 | - | 75 | 60 | |||
More Information
Set Membership: |
|

CD3D
Genomic Map