Assay Details
| Gene Symbol: CD3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034033.2 | NP_001029205.1 | 2 - 3 | 300 | 126 | |
| GenBank mRNA | BC102857.1 | - | 2 - 3 | 301 | 126 | |
More Information
Set Membership: |
|

CD3D
Genomic Map