Assay Details
| Gene Symbol: GULO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034043.2 | NP_001029215.1 | 2 - 3 | 209 | 90 | |
| GenBank mRNA | BC102936.1 | - | 2 - 3 | 210 | 90 | |
More Information
Set Membership: |
|

GULO
Genomic Map