Assay Details
| Gene Symbol: GABARAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034048.1 | NP_001029220.1 | 1 - 2 | 139 | 81 | |
| GenBank mRNA | BC103060.1 | - | 1 - 2 | 139 | 81 | |
More Information
Set Membership: |
|

GABARAP
Genomic Map