Assay Details
| Gene Symbol: GABARAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034048.1 | NP_001029220.1 | 2 - 3 | 213 | 95 | |
| GenBank mRNA | BC103060.1 | - | 2 - 3 | 213 | 95 | |
More Information
Set Membership: |
|

GABARAP
Genomic Map