Assay Details
Gene Symbol: GHITM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034052.2 | NP_001029224.1 | 3 - 4 | 317 | 70 | |
GenBank mRNA | AB098901.1 | - | 3 - 4 | 299 | 70 | |
BC102813.1 | - | 3 - 4 | 318 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
