Assay Details
| Gene Symbol: RIDA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034208.2 | NP_001029380.1 | 5 - 6 | 422 | 61 | |
| GenBank mRNA | BC102164.1 | - | 5 - 6 | 424 | 61 | |
More Information
Set Membership: |
|

RIDA
Genomic Map