Assay Details
| Gene Symbol: PAPSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034210.2 | NP_001029382.1 | 11 - 12 | 1762 | 101 | |
| GenBank mRNA | BC102372.1 | - | 11 - 12 | 1763 | 101 | |
More Information
Set Membership: |
|

PAPSS1
Genomic Map