Assay Details
| Gene Symbol: CES2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034260.2 | NP_001029432.1 | 9 - 10 | 1393 | 71 | |
| GenBank mRNA | BC102288.1 | - | 9 - 10 | 1394 | 71 | |
More Information
Set Membership: |
|

CES2
Genomic Map