Assay Details
| Gene Symbol: ARL6IP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034281.2 | NP_001029453.1 | 1 - 2 | 412 | 92 | |
| GenBank mRNA | BC105178.1 | - | 1 - 2 | 415 | 92 | |
More Information
Set Membership: |
|

ARL6IP6
Genomic Map