Assay Details
| Gene Symbol: TPPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034284.2 | NP_001029456.1 | 3 - 4 | 445 | 54 | |
| GenBank mRNA | BC102535.1 | - | 3 - 4 | 446 | 54 | |
More Information
Set Membership: |
|

TPPP2
Genomic Map