Assay Details
| Gene Symbol: CLCA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034300.1 | NP_001029472.1 | 8 - 9 | 1412 | 73 | |
| GenBank mRNA | BC103387.1 | - | 8 - 9 | 1412 | 73 | |
More Information
Set Membership: |
|

CLCA4
Genomic Map