Assay Details
| Gene Symbol: IL22RA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034311.2 | NP_001029483.1 | 5 - 6 | 692 | 63 | |
| GenBank mRNA | BC103413.1 | - | 5 - 6 | 693 | 63 | |
More Information
Set Membership: |
|

IL22RA1
Genomic Map