Assay Details
| Gene Symbol: NAGK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034314.2 | NP_001029486.1 | 4 - 5 | 404 | 59 | |
| GenBank mRNA | BC102780.1 | - | 4 - 5 | 405 | 59 | |
More Information
Set Membership: |
|

NAGK
Genomic Map