Assay Details
| Gene Symbol: AHCY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034315.1 | NP_001029487.1 | 5 - 6 | 654 | 60 | |
| GenBank mRNA | BC105194.1 | - | 5 - 6 | 654 | 60 | |
| BT030537.1 | - | 5 - 6 | 642 | 60 | ||
More Information
Set Membership: |
|

AHCY
Genomic Map