Assay Details
| Gene Symbol: DDX56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034329.2 | NP_001029501.1 | 13 - 14 | 1579 | 59 | |
| GenBank mRNA | BC103169.1 | - | 13 - 14 | 1581 | 59 | |
More Information
Set Membership: |
|

DDX56
Genomic Map