Assay Details
| Gene Symbol: SUOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034366.1 | NP_001029538.1 | 2 - 3 | 98 | 90 | |
| GenBank mRNA | BC104578.1 | - | 2 - 3 | 98 | 90 | |
More Information
Set Membership: |
|

SUOX
Genomic Map