Assay Details
| Gene Symbol: POLE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034390.1 | NP_001029562.1 | 1 - 2 | 128 | 110 | |
| GenBank mRNA | BC102772.1 | - | 1 - 2 | 128 | 110 | |
| BT030577.1 | - | 1 - 2 | 102 | 110 | ||
More Information
Set Membership: |
|

POLE3
Genomic Map