Assay Details
| Gene Symbol: POLE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034390.1 | NP_001029562.1 | 2 - 3 | 214 | 65 | |
| GenBank mRNA | BC102772.1 | - | 2 - 3 | 214 | 65 | |
| BT030577.1 | - | 2 - 3 | 188 | 65 | ||
More Information
Set Membership: |
|

POLE3
Genomic Map