Assay Details
| Gene Symbol: FXYD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034399.1 | NP_001029571.1 | 5 - 6 | 251 | 64 | |
| GenBank mRNA | BC104508.1 | - | 5 - 6 | 251 | 64 | |
More Information
Set Membership: |
|

FXYD6
Genomic Map