Assay Details
| Gene Symbol: C1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034407.1 | NP_001029579.1 | 1 - 2 | 59 | 100 | |
| GenBank mRNA | BC103405.1 | - | 1 - 2 | 59 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103405.1, NM_001034407.1 transcripts.
Set Membership: |
|

C1R
Genomic Map