Assay Details
| Gene Symbol: C1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034407.1 | NP_001029579.1 | 2 - 3 | 280 | 92 | |
| GenBank mRNA | BC103405.1 | - | 2 - 3 | 280 | 92 | |
| BT030568.1 | - | 2 - 3 | 263 | 92 | ||
More Information
Set Membership: |
|

C1R
Genomic Map