Assay Details
| Gene Symbol: POLR1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034408.2 | NP_001029580.1 | 2 - 3 | 212 | 61 | |
| GenBank mRNA | BC103202.1 | - | 2 - 3 | 213 | 61 | |
More Information
Set Membership: |
|

POLR1E
Genomic Map