Assay Details
| Gene Symbol: EBNA1BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034417.2 | NP_001029589.1 | 5 - 6 | 708 | 63 | |
| GenBank mRNA | BC102356.1 | - | 5 - 6 | 709 | 63 | |
More Information
Set Membership: |
|

EBNA1BP2
Genomic Map