Assay Details
| Gene Symbol: MAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034424.2 | NP_001029596.1 | 6 - 7 | 1002 | 80 | |
| GenBank mRNA | BC102224.1 | - | 6 - 7 | 1004 | 80 | |
More Information
Set Membership: |
|

MAF1
Genomic Map