Assay Details
| Gene Symbol: RPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034427.2 | NP_001029599.1 | 3 - 4 | 294 | 95 | |
| GenBank mRNA | BC103268.1 | - | 3 - 4 | 295 | 95 | |
| BC149500.1 | - | 1 - 2 | 2243 | 95 | ||
More Information
Set Membership: |
|

RPA3
Genomic Map