Assay Details
| Gene Symbol: CREB3L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034432.2 | NP_001029604.1 | 5 - 6 | 808 | 63 | |
| GenBank mRNA | BC103320.1 | - | 5 - 6 | 810 | 63 | |
More Information
Set Membership: |
|

CREB3L3
Genomic Map