Assay Details
| Gene Symbol: RPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034434.2 | NP_001029606.1 | 1 - 2 | 40 | 69 | |
| GenBank mRNA | BC102445.1 | - | 1 - 2 | 41 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102445.1, NM_001034434.2 transcripts.
Set Membership: |
|

RPL30
Genomic Map